Learn More
(-)-2-Azabicyclo[2.2.1]hept-5-en-3-one, 98%, > 95% ee, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 297040010
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
(-)-2-Azabicyclo[2.2.1]hept-5-en-3-one | |
79200-56-9 | |
(c=1, CH2Cl2) (1 cm cell), (c=1, CH2Cl2) (1 cm cell) at 0nm, 0.1 max. at 400nm | |
95% min. (HPLC) | |
97.5% min. (HPLC) | |
C6H7NO | |
1g | |
1r,4s-2-azabicyclo 2.2.1 hept-5-en-3-one, --1r,4s-2-azabicyclo 2.2.1 hept-5-en-3-one, 1s,4r-3-azabicyclo 2.2.1 hept-5-en-2-one, 2-azabicyclo 2,2,1 hept-5-en-3-one, vince lactam, 2-azabicyclo 2.2.1 hept-5-en-3-one, 1r,4s, --2-azabicyclo 2.2.1 hept-5-en-3-one, --vince lactam, 1s-+-2-azabicyclo 2.2.1 hept-5-en-3-one | |
C1C2C=CC1NC2=O | |
109.13 | |
109.13 |
>95%ee | |
97.5 | |
100.0 | |
Authentic | |
Glass bottle | |
MFCD00211274 | |
-560 | |
DDUFYKNOXPZZIW-UHNVWZDZSA-N | |
(1S,4R)-3-azabicyclo[2.2.1]hept-5-en-2-one | |
2725037 | |
98%,>95% ee |
Chemical Identifiers
79200-56-9 | |
109.13 | |
DDUFYKNOXPZZIW-UHNVWZDZSA-N | |
2725037 | |
C1C2C=CC1NC2=O |
C6H7NO | |
MFCD00211274 | |
1r,4s-2-azabicyclo 2.2.1 hept-5-en-3-one, --1r,4s-2-azabicyclo 2.2.1 hept-5-en-3-one, 1s,4r-3-azabicyclo 2.2.1 hept-5-en-2-one, 2-azabicyclo 2,2,1 hept-5-en-3-one, vince lactam, 2-azabicyclo 2.2.1 hept-5-en-3-one, 1r,4s, --2-azabicyclo 2.2.1 hept-5-en-3-one, --vince lactam, 1s-+-2-azabicyclo 2.2.1 hept-5-en-3-one | |
(1S,4R)-3-azabicyclo[2.2.1]hept-5-en-2-one |
Safety and Handling
GHS H Statement
Causes serious eye damage.
May cause an allergic skin reaction.
Harmful if swallowed.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
GHS Signal Word: Danger
RUO â Research Use Only