Please login to your online account to display your discounted pricing

2-Anilinopyrimidine-5-carboxylic acid, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC69201CB

 View more versions of this product

Catalog No. CC69201CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 215.21
Quantity 250mg
Assay 97%
CAS Max % 100.0
Chemical Name or Material 2-Anilinopyrimidine-5-carboxylic acid
Molecular Formula C11H9N3O2
CAS Min % 96.5
MDL Number MFCD09864415
Packaging Amber Glass Bottle
Assay Percent Range 97%
CAS 450368-25-9

Specifications

Chemical Name or Material 2-Anilinopyrimidine-5-carboxylic acid
CAS 450368-25-9
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H9N3O2
Formula Weight 215.21
MDL Number MFCD09864415