Please login to your online account to display your discounted pricing

2-Anilinopyrimidine-5-carbaldehyde, 97%, Maybridge


Quantity 250mg
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC69204CB

 View more versions of this product

Catalog No. CC69204CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 199.21
Quantity 250mg
Assay 97%
Chemical Name or Material 2-anilinopyrimidine-5-carbaldehyde
Assay Percent Range 97%
Molecular Formula C11H9N3O
Packaging Amber Glass Bottle
Health Hazard 1 GHS Signal Word: Warning
CAS 108002-87-5

Specifications

Chemical Name or Material 2-anilinopyrimidine-5-carbaldehyde
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H9N3O
Formula Weight 199.21
Accessories

GHS Signal Word: Warning

SDS & Safety Documents