Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Anilinopyridine, 97%

Chemical Identifiers

Chemical Name or Material 2-Anilinopyridine
CAS 6631-37-4
Formula Weight 170.21
MDL Number MFCD00006248
Synonym N-Phenylpyridin-2-amine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5790903

  SDS
alfa aesar™
H5790903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH57909MD

  SDS
alfa aesar™
H57909MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 170.21
Boiling Point 205° to 206°C (7mmHg)
Melting Point 106° to 112°C
CAS 6631-37-4
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Density 1.249g/mL
Physical Form Solid
Chemical Name or Material 2-Anilinopyridine
Quantity 1g
MDL Number MFCD00006248
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Synonym N-Phenylpyridin-2-amine
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.