Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Anilino-4,6-di(4-morpholinyl)-1,3,5-triazine, 97%

$392.51 - $392.51

Chemical Identifiers

Chemical Name or Material 2-Anilino-4,6-di(4-morpholinyl)-1,3,5-triazine
CAS 93438-27-8
Molecular Formula C17H22N6O2
Formula Weight 342.4
MDL Number MFCD00272422
Synonym AMMT; 2-Anilino-4,6-bis(4-morpholinyl)-1,3,5-triazine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2574103

 
alfa aesar™
B2574103
1g Each for $392.51
Description & Specifications

Specifications

Formula Weight 342.4
Chemical Name or Material 2-Anilino-4,6-di(4-morpholinyl)-1,3,5-triazine
Assay 97%
Molecular Formula C17H22N6O2
MDL Number MFCD00272422
CAS 93438-27-8
Synonym AMMT; 2-Anilino-4,6-bis(4-morpholinyl)-1,3,5-triazine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 171° to 174°C
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.