missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Aminothiophenol, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 153901000

 View more versions of this product

Catalog No. AC153901000



Specifications

Specifications

2-Aminothiophenol
79°C
Glass bottle
1.16
Yellow to Amber
100g
137-07-5
100.0
C6H7NS
MFCD00007702
1-amino-2-mercaptobenzene
VRVRGVPWCUEOGV-UHFFFAOYSA-N
2-aminobenzenethiol
8713
Liquid
1.1600g/mL
Authentic
1.6400 to 1.6440
234.0°C
20.0°C
98%
97.5
97.5% min. (Argentometry)
H2NC6H4SH
13, 397
Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
C1=CC=C(C(=C1)N)S
125.19
125.19
98%

Chemical Identifiers

137-07-5
125.19
VRVRGVPWCUEOGV-UHFFFAOYSA-N
8713
C1=CC=C(C(=C1)N)S
C6H7NS
MFCD00007702
1-amino-2-mercaptobenzene
2-aminobenzenethiol
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED

GHS Signal Word: Danger

EINECSNumber : 205-277-3

RTECSNumber : DC0600000

TSCA : TSCA

Recommended Storage : oxidized (disulphide) compound is steadily formed in air

SDS Documents