missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Aminothiazole hydrochloride, 97%

$81.57 - $371.20

Chemical Identifiers

CAS 6142-05-8
Molecular Formula C14H21NaO3S
Molecular Weight (g/mol) 292.37
MDL Number MFCD00270826
InChI Key ASJUTVUXOMPOQH-UHFFFAOYSA-M
Synonym 4-n-octylbenzenesulfonic acid sodium salt, 4-octylbenzenesulfonic acid sodium salt, acmc-2097it, dsstox_cid_27594, dsstox_gsid_47594, dsstox_rid_82441, hts5em67p9, sodium 4-n-octylbenzenesulfonate, sodium 4-octylbenzenesulfonate, unii-hts5em67p9
PubChem CID 23670838
IUPAC Name sodium;4-octylbenzenesulfonate
SMILES CCCCCCCCC1=CC=C(C=C1)S(=O)(=O)[O-].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6506822 View Documents Alfa Aesar™
H6506822
100g Each for $81.57
 
AAH6506836 View Documents Alfa Aesar™
H6506836
500g Each for $371.20
 
Specifications

Chemical Identifiers

6142-05-8
292.37
ASJUTVUXOMPOQH-UHFFFAOYSA-M
23670838
CCCCCCCCC1=CC=C(C=C1)S(=O)(=O)[O-].[Na+]
C14H21NaO3S
MFCD00270826
4-n-octylbenzenesulfonic acid sodium salt, 4-octylbenzenesulfonic acid sodium salt, acmc-2097it, dsstox_cid_27594, dsstox_gsid_47594, dsstox_rid_82441, hts5em67p9, sodium 4-n-octylbenzenesulfonate, sodium 4-octylbenzenesulfonate, unii-hts5em67p9
sodium;4-octylbenzenesulfonate

Specifications

100g
C14H21NaO3S
5404572
ASJUTVUXOMPOQH-UHFFFAOYSA-M
sodium;4-octylbenzenesulfonate
23670838
Solid
2-Aminothiazole hydrochloride
6142-05-8
MFCD00270826
4-n-octylbenzenesulfonic acid sodium salt, 4-octylbenzenesulfonic acid sodium salt, acmc-2097it, dsstox_cid_27594, dsstox_gsid_47594, dsstox_rid_82441, hts5em67p9, sodium 4-n-octylbenzenesulfonate, sodium 4-octylbenzenesulfonate, unii-hts5em67p9
CCCCCCCCC1=CC=C(C=C1)S(=O)(=O)[O-].[Na+]
292.37
136.6
97%
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 228-135-2

TSCA : No

Recommended Storage : Ambient temperatures