missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Aminothiazole, 97%

Manufacturer:  Alfa Aesar™ A1202622

 View more versions of this product

Catalog No. AAA1202622


 

Description

Description

Solubility
Soluble in water, alcohol and ether.

Notes
Incompatible with strong oxidizing agents, strong acids, acid chlorides, acid anhydrides, nitric acid and sulfuric acid.
Specifications

Specifications

2-Aminothiazole
97%
117°C (15mmHg)
88°C to 92°C
100g
RAIPHJJURHTUIC-UHFFFAOYSA-N
1,3-thiazol-2-amine
2155
100.14
96-50-4
C3H4N2S
MFCD00005325
105738
14,479
C1=CSC(=N1)N
100.139
CHEBI:40782

Chemical Identifiers

96-50-4
100.139
RAIPHJJURHTUIC-UHFFFAOYSA-N
CHEBI:40782
C3H4N2S
MFCD00005325
2155
Safety and Handling

Safety and Handling

GHS H Statement
H341-H302
Suspected of causing genetic defects.
Harmful if swallowed.

GHS P Statement
P281-P264-P301+P312-P308+P313-P405-P501a
Use personal protective equipment as required.
Wash thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : XJ2100000

TSCA : TSCA

SDS Documents