Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Aminothiazole-5-carboxaldehyde, 95%

$177.01 - $177.01

Chemical Identifiers

Chemical Name or Material 2-Aminothiazole-5-carboxaldehyde
CAS 1003-61-8
Molecular Formula C4H4N2OS
Formula Weight 128.15
MDL Number MFCD02179570
Synonym 2-Amino-5-formylthiazole
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2711203

  SDS
alfa aesar™
H2711203
1g Each for $177.01
Description & Specifications

Specifications

Formula Weight 128.15
Chemical Name or Material 2-Aminothiazole-5-carboxaldehyde
Assay 95%
Molecular Formula C4H4N2OS
Melting Point ≈160°C
MDL Number MFCD02179570
CAS 1003-61-8
Quantity 1g
Synonym 2-Amino-5-formylthiazole
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.