Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Aminothiazole-4-carboxamide, 95%

Chemical Identifiers

Chemical Name or Material 2-Aminothiazole-4-carboxamide
CAS 118452-02-1
Formula Weight 143.16
MDL Number MFCD08689593
Beilstein 4969494
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6655703

 
alfa aesar™
H6655703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6655706

 
alfa aesar™
H6655706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66557MD

 
alfa aesar™
H66557MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 143.16
CAS 118452-02-1
MDL Number MFCD08689593
Assay 95%
Beilstein 4969494
Physical Form Solid
Quantity 1g
Chemical Name or Material 2-Aminothiazole-4-carboxamide