Learn More
2-Aminoterephthalic acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2444122
Description
2-Aminoterephthalic acid is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Aminoterephthalic acid is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
Specifications
2-Aminoterephthalic acid | |
>300°C | |
C8H5NO4 | |
100 g | |
Slightly soluble in water. | |
NC1=CC(=CC=C1C([O-])=O)C([O-])=O | |
179.13 | |
181.15 |
10312-55-7 | |
Odorless | |
MFCD00134536 | |
aminoterephthalic acid, 2-aminobenzene-1,4-dicarboxylic acid, 2-amino-1,4-benzenedicarboxylic acid, 4-carboxyanthranilic acid, 1,4-benzenedicarboxylic acid, 2-amino, 2-aminoterephthalic, aminoterephthalicacid, 2,5-dicarboxyaniline, 2-aminoterephtalic acid, timtec-bb sbb006751 | |
GPNNOCMCNFXRAO-UHFFFAOYSA-L | |
2-aminoterephthalic acid | |
2724822 | |
99% |
Chemical Identifiers
10312-55-7 | |
179.13 | |
GPNNOCMCNFXRAO-UHFFFAOYSA-L | |
2724822 | |
NC1=CC(=CC=C1C([O-])=O)C([O-])=O |
C8H5NO4 | |
MFCD00134536 | |
aminoterephthalic acid, 2-aminobenzene-1,4-dicarboxylic acid, 2-amino-1,4-benzenedicarboxylic acid, 4-carboxyanthranilic acid, 1,4-benzenedicarboxylic acid, 2-amino, 2-aminoterephthalic, aminoterephthalicacid, 2,5-dicarboxyaniline, 2-aminoterephtalic acid, timtec-bb sbb006751 | |
2-aminoterephthalic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only