missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Aminopyrimidine-5-carboxaldehyde, 97%

$83.58 - $993.24

Chemical Identifiers

CAS 120747-84-4
Molecular Formula C5H5N3O
Molecular Weight (g/mol) 123.115
MDL Number MFCD03701714
InChI Key DPOYRRAYGKTRAU-UHFFFAOYSA-N
Synonym 2-amino-5-formylpyrimidine
PubChem CID 769097
IUPAC Name 2-aminopyrimidine-5-carbaldehyde
SMILES C1=C(C=NC(=N1)N)C=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH54440MD View Documents Alfa Aesar™
H54440MD
250mg Each for $83.58
 
AAH5444003 View Documents Alfa Aesar™
H5444003
1g Each for $249.12
 
AAH5444006 View Documents Alfa Aesar™
H5444006
5g Each for $993.24
 
Specifications

Chemical Identifiers

120747-84-4
123.115
DPOYRRAYGKTRAU-UHFFFAOYSA-N
769097
C1=C(C=NC(=N1)N)C=O
C5H5N3O
MFCD03701714
2-amino-5-formylpyrimidine
2-aminopyrimidine-5-carbaldehyde

Specifications

250mg
C5H5N3O
2-amino-5-formylpyrimidine
C1=C(C=NC(=N1)N)C=O
123.115
123.11
2-Aminopyrimidine-5-carboxaldehyde
120747-84-4
MFCD03701714
DPOYRRAYGKTRAU-UHFFFAOYSA-N
2-aminopyrimidine-5-carbaldehyde
769097
97%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures