missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Aminopyrimidine-5-carboxaldehyde, 97%

Manufacturer:  Alfa Aesar™ H54440MD

 View more versions of this product

Catalog No. AAH54440MD



Description

Description

Specifications

Specifications

2-Aminopyrimidine-5-carboxaldehyde
120747-84-4
MFCD03701714
DPOYRRAYGKTRAU-UHFFFAOYSA-N
2-aminopyrimidine-5-carbaldehyde
769097
97%
250mg
C5H5N3O
2-amino-5-formylpyrimidine, 2-amino-5-pyrimidinecarbaldehyde, 2-amino-5-pyrimidinecarboxaldehyde, 2-amino-5-pyrimidinecarboxyaldehyde, 2-amino-pyrimidine-5-carbaldehyde, 2-aminopyrimidine-5-carboxaldehyde, 5-pyrimidinecarboxaldehyde, 2-amino, 5-pyrimidinecarboxaldehyde,2-amino, acmc-20a0p5, zlchem 786
C1=C(C=NC(=N1)N)C=O
123.115
123.11

Chemical Identifiers

120747-84-4
123.115
DPOYRRAYGKTRAU-UHFFFAOYSA-N
769097
C1=C(C=NC(=N1)N)C=O
C5H5N3O
MFCD03701714
2-amino-5-formylpyrimidine, 2-amino-5-pyrimidinecarbaldehyde, 2-amino-5-pyrimidinecarboxaldehyde, 2-amino-5-pyrimidinecarboxyaldehyde, 2-amino-pyrimidine-5-carbaldehyde, 2-aminopyrimidine-5-carboxaldehyde, 5-pyrimidinecarboxaldehyde, 2-amino, 5-pyrimidinecarboxaldehyde,2-amino, acmc-20a0p5, zlchem 786
2-aminopyrimidine-5-carbaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents