Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Aminopyrimidine-5-boronic acid, 95%

Chemical Identifiers

Chemical Name or Material 2-Aminopyrimidine-5-boronic acid
CAS 936250-22-5
Formula Weight 138.92
MDL Number MFCD07375147
Beilstein 15758675
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6640303

  SDS
alfa aesar™
H6640303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6640306

  SDS
alfa aesar™
H6640306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66403MD

  SDS
alfa aesar™
H66403MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 138.92
Melting Point 206° to 211°C
Chemical Name or Material 2-Aminopyrimidine-5-boronic acid
CAS 936250-22-5
Assay 95%
Physical Form Solid
Beilstein 15758675
MDL Number MFCD07375147
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Storage Note 1 2° to 8°C
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
2-Aminopyrimidine-5-boronic acid is used as pharmaceutical intermediates.

Notes
Store at 4°C. Protect from heat. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C