Learn More
2-Aminopyridine, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1237430
Description
A compound has been shown to reversibly block voltage-dependent potassium channels. Also a common impurity from the synthesis of compounds found in hair dye. Used as a water miscible solvent and as an intermediate in manufacturing pharmaceuticals like diuretics. It is used as a research tool, in characterizing subtypes of the potassium channel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA compound has been shown to reversibly block voltage-dependent potassium channels. Also a common impurity from the synthesis of compounds found in hair dye. Used as a water miscible solvent and as an intermediate in manufacturing pharmaceuticals like diuretics. It is used as a research tool, in characterizing subtypes of the potassium channel.
Solubility
Slightly soluble in water(8g/100 mL at 19°C)
Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids.
Specifications
2-Aminopyridine | |
56°C to 60°C | |
92°C (197°F) | |
MFCD00006312 | |
UN2671 | |
Hygroscopic | |
2-aminopyridine, 2-pyridinamine, 2-pyridylamine, aminopyridine, pyridinamine, o-aminopyridine, 2-aminopyridin, 2-aminopryidine, amino-2 pyridine, pyridine, 2-amino | |
ICSNLGPSRYBMBD-UHFFFAOYSA-N | |
pyridin-2-amine | |
10439 | |
99% |
504-29-0 | |
209°C to 211°C | |
C5H6N2 | |
250 g | |
105785 | |
14,473 | |
Slightly soluble in water(8g/100 mL at 19°C) | |
C1=CC=NC(=C1)N | |
94.117 | |
94.12 |
Chemical Identifiers
504-29-0 | |
94.117 | |
ICSNLGPSRYBMBD-UHFFFAOYSA-N | |
10439 | |
C1=CC=NC(=C1)N |
C5H6N2 | |
MFCD00006312 | |
2-aminopyridine, 2-pyridinamine, 2-pyridylamine, aminopyridine, pyridinamine, o-aminopyridine, 2-aminopyridin, 2-aminopryidine, amino-2 pyridine, pyridine, 2-amino | |
pyridin-2-amine |
Safety and Handling
GHS H Statement
H301-H331-H312-H315-H319-H335
Toxic if swallowed.
Toxic if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H312-H315-H319-H500
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: AMINOPYRIDINES
EINECSNumber : 207-988-4
RTECSNumber : US1575000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only