Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Aminopyridine-4-methanol, 97%

$151.00 - $151.00

Chemical Identifiers

Chemical Name or Material 2-Aminopyridine-4-methanol
CAS 105250-17-7
Molecular Formula C6H8N2O
Formula Weight 124.14
MDL Number MFCD03791261
Beilstein 8760550
Synonym 2-Amino-4-(hydroxymethyl)pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1984603

  SDS
alfa aesar™
L1984603
1g Each for $151.00
Description & Specifications

Specifications

Formula Weight 124.14
Chemical Name or Material 2-Aminopyridine-4-methanol
Melting Point 81° to 85°C
Quantity 1g
Assay 97%
Molecular Formula C6H8N2O
MDL Number MFCD03791261
CAS 105250-17-7
Synonym 2-Amino-4-(hydroxymethyl)pyridine
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Recommended Storage 2° to 8°C
Beilstein 8760550
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
2-Aminopyridine-4-methanol is used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C