missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Aminopyridine-4-carboxylic acid, 96%

$58.95 - $758.43

Chemical Identifiers

CAS 13362-28-2
Molecular Formula C6H6N2O2
Formula Weight 138.13
MDL Number MFCD00233714
Beilstein 3156
Synonym 2-Aminoisonicotinic acid
Chemical Name or Material 2-Aminopyridine-4-carboxylic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1984503 SDS Alfa Aesar™
L1984503
1g Each for $58.95
AAL1984506 SDS Alfa Aesar™
L1984506
5g Each for $191.47
AAL1984514 SDS Alfa Aesar™
L1984514
25g Each for $758.43
Specifications Description & Specifications

Chemical Identifiers

13362-28-2
138.13
3156
2-Aminopyridine-4-carboxylic acid
C6H6N2O2
MFCD00233714
2-Aminoisonicotinic acid

Specifications

13362-28-2
1g
C6H6N2O2
MFCD00233714
2-Aminoisonicotinic acid
96%
300° to 302°C
Odorless
138.13
3156
2-Aminopyridine-4-carboxylic acid

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning