Learn More
2-Aminopyridine-3-carbonitrile, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC441430050
Specifications
2-Aminopyridine-3-carbonitrile | |
Glass bottle | |
130°C to 136°C | |
24517-64-4 | |
100.0 | |
C6H5N3 | |
YYXDQRRDNPRJFL-UHFFFAOYSA-N | |
2-aminopyridine-3-carbonitrile | |
818259 | |
Crystalline Powder |
Authentic | |
White to Brown | |
5g | |
96.0 | |
97% | |
2-amino-3-cyanopyridine, 2-amino-3-pyridinecarbonitrile, 2-amino-nicotinonitrile, 2-amino-pyridine-3-carbonitrile, 2-aminonicotinonitrile, 3-cyanopyridin-2-amine, 3-cyanopyridine-2-amine, 3-pyridinecarbonitrile, 2-amino, aminonicotinonitrile, zlchem 506 | |
C1=CC(=C(N=C1)N)C#N | |
119.127 | |
119.13 | |
≥96.0% |
Chemical Identifiers
24517-64-4 | |
119.127 | |
2-amino-3-cyanopyridine, 2-amino-3-pyridinecarbonitrile, 2-amino-nicotinonitrile, 2-amino-pyridine-3-carbonitrile, 2-aminonicotinonitrile, 3-cyanopyridin-2-amine, 3-cyanopyridine-2-amine, 3-pyridinecarbonitrile, 2-amino, aminonicotinonitrile, zlchem 506 | |
2-aminopyridine-3-carbonitrile |
C6H5N3 | |
YYXDQRRDNPRJFL-UHFFFAOYSA-N | |
818259 | |
C1=CC(=C(N=C1)N)C#N |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning