missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Aminopyrazine, 99%

$62.97 - $421.85

Chemical Identifiers

CAS 5049-61-6
Molecular Formula C4H5N3
Formula Weight 95.11
MDL Number MFCD00006137
Beilstein 107025
Synonym Pyrazinamine
Chemical Name or Material 2-Aminopyrazine
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1305206 SDS Alfa Aesar™
A1305206
5g Each for $62.97
AAA1305214 SDS Alfa Aesar™
A1305214
25g Each for $225.88
AAA1305218 SDS Alfa Aesar™
A1305218
50g Each for $421.85
Specifications

Description

Applications
2-Aminopyrazine acts as a raw material in a four-component synthesis of imidazolidines. It is also used as an intermediate in synthetic chemistry.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
Description & Specifications

Chemical Identifiers

5049-61-6
95.11
107025
2-Aminopyrazine
C4H5N3
MFCD00006137
Pyrazinamine

Specifications

5049-61-6
5g
95.11
107025
2-Aminopyrazine
118° to 121°C
C4H5N3
MFCD00006137
Pyrazinamine
99%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning