missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Aminophenylboronic acid hydrochloride, 97%, Thermo Scientific™
Manufacturer: Thermo Fisher Scientific AC445310050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Aminophenylboronic acid hydrochloride | |
96% min. (HPLC) | |
Glass Bottle | |
5g | |
B(C1=CC=CC=C1N)(O)O.Cl | |
173.41 | |
173.41 |
863753-30-4 | |
C6H8BNO2·ClH | |
2-2-aminophenyl boronic acid hydrochloride, 2-aminophenyl boronic acid hydrochloride, 2-aminophenylboronic acid hcl, 2-aminophenylboronic acid hydrochloride, 2-aminophenylboronic acid, hcl, 2-aminophenylboronic acid, hydrochloride, 2-aminophenylboronic acid,hcl, boronic acid, 2-aminophenyl-, hydrochloride, ksc914i9f, pubchem1746 | |
WPDASZCYRKGSTO-UHFFFAOYSA-N | |
(2-aminophenyl)boronic acid;hydrochloride | |
16427090 | |
97% |
Chemical Identifiers
863753-30-4 | |
173.41 | |
2-2-aminophenyl boronic acid hydrochloride, 2-aminophenyl boronic acid hydrochloride, 2-aminophenylboronic acid hcl, 2-aminophenylboronic acid hydrochloride, 2-aminophenylboronic acid, hcl, 2-aminophenylboronic acid, hydrochloride, 2-aminophenylboronic acid,hcl, boronic acid, 2-aminophenyl-, hydrochloride, ksc914i9f, pubchem1746 | |
(2-aminophenyl)boronic acid;hydrochloride |
C6H8BNO2·ClH | |
WPDASZCYRKGSTO-UHFFFAOYSA-N | |
16427090 | |
B(C1=CC=CC=C1N)(O)O.Cl |
Safety and Handling
GHS Signal Word: Warning
RUO – Research Use Only