missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Aminophenol, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 104261000

 View more versions of this product

Catalog No. AC104261000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Aminophenol
95-55-6
100.0
White to Brown
168°C
98.5% min. (GC)
C6H7NO
MFCD00007690
13, 354
o-aminophenol, o-hydroxyaniline, 2-hydroxyaniline, phenol, 2-amino, 2-amino-1-hydroxybenzene, fouramine op, benzofur gg, pelagol grey gg, pelagol 3ga, nako yellow 3ga
CDAWCLOXVUBKRW-UHFFFAOYSA-N
2-aminophenol
5801
109.13
Powder
99%
98.5
172°C to 177°C
164°C (15.0 Mbar)
Authentic
Glass bottle
H2NC6H4OH
100 g
15, 457
Solubility in water: 17g/L (20°C). Other solubilities: soluble in ether,1g in 23 mL alcohol,slightly soluble in benzene
C1=CC=C(C(=C1)N)O
109.13
CHEBI:18112
99%

Chemical Identifiers

95-55-6
109.13
CDAWCLOXVUBKRW-UHFFFAOYSA-N
5801
2-aminophenol
C6H7NO
MFCD00007690
o-aminophenol, o-hydroxyaniline, 2-hydroxyaniline, phenol, 2-amino, 2-amino-1-hydroxybenzene, fouramine op, benzofur gg, pelagol grey gg, pelagol 3ga, nako yellow 3ga
CHEBI:18112
C1=CC=C(C(=C1)N)O
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Suspected of causing genetic defects.
May cause an allergic skin reaction.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN:

GHS Signal Word: Warning

EINECSNumber : 202-431-1

RTECSNumber : SJ4950000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only