missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Aminophenol, 99%, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific 104265000

 View more versions of this product

Catalog No. AC104265000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

2-Aminophenol
Authentic
164°C (15.0 mbar)
172°C to 177°C
99%
98.5
98.5% min. (GC)
H2NC6H4OH
13, 354
2-amino-1-hydroxybenzene, 2-hydroxyaniline, benzofur gg, fouramine op, nako yellow 3ga, o-aminophenol, o-hydroxyaniline, pelagol 3ga, pelagol grey gg, phenol, 2-amino
CDAWCLOXVUBKRW-UHFFFAOYSA-N
2-aminophenol
5801
109.13
99%
168°C
Glass bottle
White to Brown
500g
95-55-6
100.0
C6H7NO
MFCD00007690
15, 457
Solubility in water: 17g/L (20°C). Other solubilities: soluble in ether,1g in 23 mL alcohol,slightly soluble in benzene
C1=CC=C(C(=C1)N)O
109.13
CHEBI:18112
Powder

Chemical Identifiers

95-55-6
109.13
CDAWCLOXVUBKRW-UHFFFAOYSA-N
5801
2-aminophenol
C6H7NO
MFCD00007690
2-amino-1-hydroxybenzene, 2-hydroxyaniline, benzofur gg, fouramine op, nako yellow 3ga, o-aminophenol, o-hydroxyaniline, pelagol 3ga, pelagol grey gg, phenol, 2-amino
CHEBI:18112
C1=CC=C(C(=C1)N)O
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
Suspected of causing genetic defects.
May cause an allergic skin reaction.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN:

GHS Signal Word: Warning

EINECSNumber : 202-431-1

RTECSNumber : SJ4950000

TSCA : TSCA

SDS Documents
Product Certifications

RUO – Research Use Only