Please login to your online account to display your discounted pricing

2-Aminonicotinic Acid 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  104160250

 View more versions of this product

Catalog No. AC104160250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 138.13
Quantity 25g
Color Off-white to light yellow
EINECS Number 226-296-3
TSCA TSCA
Beilstein 22, 542
CAS Max % 100.0
Name Note 98%
MDL Number MFCD00006318
CAS 5345-47-1
Synonym 2-Aminopyridine-3-carboxylic acid
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2-Aminonicotinic acid
Assay Percent Range 98%
Molecular Formula C6H6N2O2
Linear Formula H2N(C5H3N)COOH
CAS Min % 97.5
Infrared Spectrum Authentic
Packaging Glass bottle

Specifications

Chemical Name or Material 2-Aminonicotinic acid
Name Note 98%
CAS 5345-47-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H6N2O2
Linear Formula H2N(C5H3N)COOH
Formula Weight 138.13
MDL Number MFCD00006318
Synonym 2-Aminopyridine-3-carboxylic acid
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents