Please login to your online account to display your discounted pricing

2-Aminonicotinic Acid 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  104160050

 View more versions of this product

Catalog No. AC104160050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 138.13
Physical Form Crystalline Powder
Quantity 5g
Color Off-white to light yellow
Beilstein 22, 542
TSCA TSCA
EINECS Number 226-296-3
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 98%
Molecular Formula C6H6N2O2
MDL Number MFCD00006318
CAS 5345-47-1
Chemical Name or Material 2-Aminonicotinic acid
Linear Formula H2N(C5H3N)COOH
Assay Percent Range 98%
CAS Min % 97.5
Packaging Glass bottle
Synonym 2-Aminopyridine-3-carboxylic acid

Specifications

Chemical Name or Material 2-Aminonicotinic acid
Name Note 98%
CAS 5345-47-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H6N2O2
Linear Formula H2N(C5H3N)COOH
Formula Weight 138.13
MDL Number MFCD00006318
Synonym 2-Aminopyridine-3-carboxylic acid
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents