missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Aminomethyl-5-fluorobenzeneboronic acid hydrochloride, 95%

Manufacturer:  Alfa Aesar™ H53097MD

 View more versions of this product

Catalog No. AAH53097MD



Specifications

Specifications

2-Aminomethyl-5-fluorobenzeneboronic acid hydrochloride
250mg
C7H10BClFNO2
2-aminomethyl-5-fluorophenyl boronic acid hydrochloride, 2-aminomethyl-5-fluorophenylboronic acid, hcl, 2-aminomethyl-5-fluoro phenylboronic acid hydrochloride, 2-borono-4-fluorobenzylamine hydrochloride, 2-aminomethyl-5-fluorophenylboronic acid hydrochloride, 2-aminomethyl-5-fluorophenyl boronic acid hydrochloride, 2-borono-4-fluorobenzylamine, hcl, 2-aminomethyl-5-fluorophenylboronic acid hcl, 2-aminomethyl-5-fluorophenylboronic acid hydrochloride, 2-aminomethyl-5-fluorophenyl boronicacidhydrochloride
B(C1=C(C=CC(=C1)F)CN)(O)O.Cl
205.42
205.42
152°C to 158°C
850568-03-5
MFCD04115666
URBGTEWFXWJWPS-UHFFFAOYSA-N
[2-(aminomethyl)-5-fluorophenyl]boronic acid;hydrochloride
2782819
95%

Chemical Identifiers

850568-03-5
205.42
URBGTEWFXWJWPS-UHFFFAOYSA-N
2782819
B(C1=C(C=CC(=C1)F)CN)(O)O.Cl
C7H10BClFNO2
MFCD04115666
2-aminomethyl-5-fluorophenyl boronic acid hydrochloride, 2-aminomethyl-5-fluorophenylboronic acid, hcl, 2-aminomethyl-5-fluoro phenylboronic acid hydrochloride, 2-borono-4-fluorobenzylamine hydrochloride, 2-aminomethyl-5-fluorophenylboronic acid hydrochloride, 2-aminomethyl-5-fluorophenyl boronic acid hydrochloride, 2-borono-4-fluorobenzylamine, hcl, 2-aminomethyl-5-fluorophenylboronic acid hcl, 2-aminomethyl-5-fluorophenylboronic acid hydrochloride, 2-aminomethyl-5-fluorophenyl boronicacidhydrochloride
[2-(aminomethyl)-5-fluorophenyl]boronic acid;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents