missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(Aminomethyl)-1-ethylpyrrolidine, 95%, ACROS Organics™
$174.32 - $571.62
Chemical Identifiers
CAS | 26116-12-1 |
---|---|
Molecular Formula | C7H16N2 |
Molecular Weight (g/mol) | 128.22 |
MDL Number | MFCD00003178 |
InChI Key | UNRBEYYLYRXYCG-UHFFFAOYSA-N |
Synonym | 1-1-ethylpyrrolidin-2-yl methanamine, 1-ethyl-2-aminomethyl pyrrolidine, 1-ethyl-2-aminomethylpyrrolidine, 1-ethylpyrrolidin-2-yl methanamine, 1-ethylpyrrolidin-2-yl methylamine, 1-ethylpyrrolidin-2-ylmethylamine, 2-aminomethyl-1-ethylpyrrolidine, 2-pyrrolidinemethanamine, 1-ethyl, n-ethyl-2-aminomethylpyrrolidine |
PubChem CID | 117295 |
IUPAC Name | (1-ethylpyrrolidin-2-yl)methanamine |
SMILES | CCN1CCCC1CN |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC179480100
|
Thermo Fisher Scientific
179480100 |
10g | Glass bottle |
Each for $174.32
|
|
AC179480500
|
Thermo Fisher Scientific
179480500 |
50g | Glass bottle |
Each for $571.62
|
|
Chemical Identifiers
26116-12-1 | |
128.22 | |
UNRBEYYLYRXYCG-UHFFFAOYSA-N | |
117295 | |
CCN1CCCC1CN |
C7H16N2 | |
MFCD00003178 | |
1-1-ethylpyrrolidin-2-yl methanamine, 1-ethyl-2-aminomethyl pyrrolidine, 1-ethyl-2-aminomethylpyrrolidine, 1-ethylpyrrolidin-2-yl methanamine, 1-ethylpyrrolidin-2-yl methylamine, 1-ethylpyrrolidin-2-ylmethylamine, 2-aminomethyl-1-ethylpyrrolidine, 2-pyrrolidinemethanamine, 1-ethyl, n-ethyl-2-aminomethylpyrrolidine | |
(1-ethylpyrrolidin-2-yl)methanamine |
Specifications
0.8840g/mL | |
Authentic | |
1.4655 to 1.4675 | |
58.0°C to 60.0°C (16.0mmHg) | |
10g | |
94.0 | |
95% | |
MFCD00003178 | |
UNRBEYYLYRXYCG-UHFFFAOYSA-N | |
(1-ethylpyrrolidin-2-yl)methanamine | |
117295 | |
Liquid | |
2-(Aminomethyl)-1-ethylpyrrolidine, 95% |
56°C | |
Glass bottle | |
0.884 | |
Yellow | |
26116-12-1 | |
100.0 | |
C7H16N2 | |
1-1-ethylpyrrolidin-2-yl methanamine, 1-ethyl-2-aminomethyl pyrrolidine, 1-ethyl-2-aminomethylpyrrolidine, 1-ethylpyrrolidin-2-yl methanamine, 1-ethylpyrrolidin-2-yl methylamine, 1-ethylpyrrolidin-2-ylmethylamine, 2-aminomethyl-1-ethylpyrrolidine, 2-pyrrolidinemethanamine, 1-ethyl, n-ethyl-2-aminomethylpyrrolidine | |
CCN1CCCC1CN | |
128.22 | |
128.22 | |
95% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p
GHS Signal Word: Danger
EINECSNumber : 247-466-3
TSCA : TSCA