missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Aminomethyl)-1-ethylpyrrolidine, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 179480100

 View more versions of this product

Catalog No. AC179480100



Description

Description

Specifications

Specifications

2-(Aminomethyl)-1-ethylpyrrolidine
56°C
Glass bottle
0.884
Yellow
95%
94.0
95%
MFCD00003178
UNRBEYYLYRXYCG-UHFFFAOYSA-N
(1-ethylpyrrolidin-2-yl)methanamine
117295
Liquid
0.8840g/mL
Authentic
1.4655 to 1.4675
58.0°C to 60.0°C (16.0mmHg)
10g
26116-12-1
100.0
C7H16N2
1-1-ethylpyrrolidin-2-yl methanamine, 1-ethyl-2-aminomethyl pyrrolidine, 1-ethyl-2-aminomethylpyrrolidine, 1-ethylpyrrolidin-2-yl methanamine, 1-ethylpyrrolidin-2-yl methylamine, 1-ethylpyrrolidin-2-ylmethylamine, 2-aminomethyl-1-ethylpyrrolidine, 2-pyrrolidinemethanamine, 1-ethyl, n-ethyl-2-aminomethylpyrrolidine
CCN1CCCC1CN
128.22
128.22
95%

Chemical Identifiers

26116-12-1
128.22
UNRBEYYLYRXYCG-UHFFFAOYSA-N
117295
CCN1CCCC1CN
C7H16N2
MFCD00003178
1-1-ethylpyrrolidin-2-yl methanamine, 1-ethyl-2-aminomethyl pyrrolidine, 1-ethyl-2-aminomethylpyrrolidine, 1-ethylpyrrolidin-2-yl methanamine, 1-ethylpyrrolidin-2-yl methylamine, 1-ethylpyrrolidin-2-ylmethylamine, 2-aminomethyl-1-ethylpyrrolidine, 2-pyrrolidinemethanamine, 1-ethyl, n-ethyl-2-aminomethylpyrrolidine
(1-ethylpyrrolidin-2-yl)methanamine
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

GHS Signal Word: Danger

EINECSNumber : 247-466-3

TSCA : TSCA

SDS Documents