missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Aminomethyl-1-Boc-piperidine, 95%

Manufacturer:  Alfa Aesar™ H5450706

 View more versions of this product

Catalog No. AAH5450706



Description

Description

Applications
Reactant for synthesis of: TRPV1 antagonists, CHK1 inhibitors, p38a MAP kinase inhibitors, Carbamoylphosphonates, Macrolactams via carbonylation/intramolecular amidation, Intramolecular transamidation.

Solubility
Soluble in DMSO.

Notes
Refrigerated (2-8°C). Incompatible with oxidizing agents.
Specifications

Specifications

2-Aminomethyl-1-Boc-piperidine
5g
C11H22N2O2
UN3082
PTVRCUVHYMGECC-UHFFFAOYSA-N
tert-butyl 2-(aminomethyl)piperidine-1-carboxylate
2756482
95%
>110°C (230°F)
370069-31-1
MFCD01317791
1-boc-2-aminomethyl piperidine
CC(C)(C)OC(=O)N1CCCCC1CN
214.309
214.31

Chemical Identifiers

370069-31-1
214.309
PTVRCUVHYMGECC-UHFFFAOYSA-N
2756482
CC(C)(C)OC(=O)N1CCCCC1CN
C11H22N2O2
MFCD01317791
1-boc-2-aminomethyl piperidine
tert-butyl 2-(aminomethyl)piperidine-1-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

Documents