Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Aminoindane, 97%

Chemical Identifiers

Chemical Name or Material 2-Aminoindane
CAS 2975-41-9
Molecular Formula C9H11N
Formula Weight 133.19g/mol
MDL Number MFCD00082597
UN Number UN2922
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3102306

 
alfa aesar™
H31023-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 229°C
Quantity 5g
Flash Point 100°C (212°F)
Odor Amine-like
Chemical Name or Material 2-Aminoindane
CAS 2975-41-9
Assay 97%
Molecular Formula C9H11N
Formula Weight 133.19g/mol
MDL Number MFCD00082597
UN Number UN2922
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

Applications
2-Aminoindan is an analgesic with sympathomimetic activity, It is an inhibitor norepinephrine methyltransferase.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.