Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-2-Aminoheptane sulfate, 99%

$372.18 - $372.18

Chemical Identifiers

Chemical Name or Material (+/-)-2-Aminoheptane sulfate
CAS 6411-75-2
Molecular Formula C14H34N2·H2SO4
Formula Weight 328.51
MDL Number MFCD00050844
Beilstein 3703311
Merck Index 14,9802
Synonym (+/-)-2-Heptylamine sulfate; (+/-)-1-Methylhexylamine sulfate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0859622

  SDS
alfa aesar™
L0859622
100g Each for $372.18
Description & Specifications

Specifications

Formula Weight 328.51
Chemical Name or Material (+/-)-2-Aminoheptane sulfate
CAS 6411-75-2
Assay 99%
Molecular Formula C14H34N2·H2SO4
Melting Point 289° to 291°C
MDL Number MFCD00050844
Quantity 100g
Beilstein 3703311
Merck Index 14,9802
Synonym (+/-)-2-Heptylamine sulfate; (+/-)-1-Methylhexylamine sulfate
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number MQ5600000
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.