Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-2-Aminobutane, 98%

Chemical Identifiers

Chemical Name or Material (+/-)-2-Aminobutane
CAS 13952-84-6
Molecular Formula C4H11N
Formula Weight 73.14
MDL Number MFCD00008094
UN Number UN2733
Beilstein 1361345
Merck Index 14,1544
Synonym (+/-)-2-Butylamine; sec-Butylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA130140F

  SDS
alfa aesar™
A130140F
2500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA13014AP

  SDS
alfa aesar™
A13014AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 73.14
TSCA TSCA
Chemical Name or Material (+/-)-2-Aminobutane
Assay 98%
Odor Amine-like
Quantity 2500mL
CAS 13952-84-6
Health Hazard 1 Danger
RTECS Number EO3325000
Merck Index 14,1544
UN Number UN2733
MDL Number MFCD00008094
Molecular Formula C4H11N
pH 12.5
Health Hazard 2 GHS H Statement
H225-H314-H318-H302-H332
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.
Flash Point −19°C (−2°F)
Beilstein 1361345
Synonym (+/-)-2-Butylamine; sec-Butylamine
Boiling Point 63° to 64°C
Melting Point −104°C
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

Applications
(+/-)-2-Aminobutane is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. It is sensitive air and light. Incompatible with strong oxidizing agents. Incompatible with strong oxidizing agents, strong acids and carbon dioxide.

Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H314-H318-H302-H332
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful if inhaled.