missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Aminobenzophenone 98%, ACROS Organics™

Manufacturer:  Acros Organics 103161000

 View more versions of this product

Catalog No. AC103161000



Specifications

Specifications

2-Aminobenzophenone
Authentic
Golden-Yellow
100g
2835-77-0
100.0
C13H11NO
MFCD00007713
2-amino-phenyl-phenyl-methanone
C1=CC=C(C=C1)C(=O)C2=CC=CC=C2N
197.237
197.24
98%
240°C
Glass bottle
105.0°C to 108.0°C
98%
97.5
98%
H2NC6H4COC6H5
14, 76
MAOBFOXLCJIFLV-UHFFFAOYSA-N
(2-aminophenyl)-phenylmethanone
76080
Fine Crystalline Powder

Chemical Identifiers

2835-77-0
197.237
MAOBFOXLCJIFLV-UHFFFAOYSA-N
76080
C1=CC=C(C=C1)C(=O)C2=CC=CC=C2N
C13H11NO
MFCD00007713
2-amino-phenyl-phenyl-methanone
(2-aminophenyl)-phenylmethanone
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 220-613-9

SDS Documents