missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Aminobenzeneboronic acid pinacol ester hydrochloride, 96%, Thermo Scientific™
$265.79 - $265.79
Chemical Identifiers
CAS | 393877-09-3 |
---|---|
Molecular Formula | C12H19BClNO2 |
Molecular Weight (g/mol) | 255.549 |
MDL Number | MFCD02179449 |
InChI Key | XWILEMYKORLIPW-UHFFFAOYSA-N |
Synonym | 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline hydrochloride, 2-aminophenylboronic acid, pinacol ester, hcl, 2-tetramethyl-1,3,2-dioxaborolan-2-yl aniline hydrochloride, 2-aminobenzeneboronic acid pinacol ester hci, 2-aminophenyl boronic acid, pinacol ester hydrochloride, 2-aminophenyl boronic acid,pinacol ester hydrochloride, 2-aminobenzeneboronic acid pinacol ester hydrochloride, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline-hydrogen chloride 1/1, benzenamine, 2-4,4,5,5-tetremethyl-1,3,2-dioxaborolan-2-yl-, hcl salt |
PubChem CID | 44118691 |
IUPAC Name | 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline;hydrochloride |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2N.Cl |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH53194MD
|
Thermo Scientific Chemicals
H53194MD |
250 mg |
Each for $265.79
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
393877-09-3 | |
255.549 | |
XWILEMYKORLIPW-UHFFFAOYSA-N | |
44118691 | |
B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2N.Cl |
C12H19BClNO2 | |
MFCD02179449 | |
2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline hydrochloride, 2-aminophenylboronic acid, pinacol ester, hcl, 2-tetramethyl-1,3,2-dioxaborolan-2-yl aniline hydrochloride, 2-aminobenzeneboronic acid pinacol ester hci, 2-aminophenyl boronic acid, pinacol ester hydrochloride, 2-aminophenyl boronic acid,pinacol ester hydrochloride, 2-aminobenzeneboronic acid pinacol ester hydrochloride, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline-hydrogen chloride 1/1, benzenamine, 2-4,4,5,5-tetremethyl-1,3,2-dioxaborolan-2-yl-, hcl salt | |
2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline;hydrochloride |
Specifications
393877-09-3 | |
C12H19BClNO2 | |
250 mg | |
XWILEMYKORLIPW-UHFFFAOYSA-N | |
2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline;hydrochloride | |
44118691 | |
96% |
161°C to 163°C | |
MFCD02179449 | |
2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline hydrochloride, 2-aminophenylboronic acid, pinacol ester, hcl, 2-tetramethyl-1,3,2-dioxaborolan-2-yl aniline hydrochloride, 2-aminobenzeneboronic acid pinacol ester hci, 2-aminophenyl boronic acid, pinacol ester hydrochloride, 2-aminophenyl boronic acid,pinacol ester hydrochloride, 2-aminobenzeneboronic acid pinacol ester hydrochloride, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline-hydrogen chloride 1/1, benzenamine, 2-4,4,5,5-tetremethyl-1,3,2-dioxaborolan-2-yl-, hcl salt | |
B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2N.Cl | |
255.549 | |
255.55 | |
2-Aminobenzeneboronic acid pinacol ester hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures