missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Aminobenzeneboronic acid pinacol ester hydrochloride, 96%

Manufacturer:  Alfa Aesar™ H5319403

 View more versions of this product

Catalog No. AAH5319403



Specifications

Specifications

2-Aminobenzeneboronic acid pinacol ester hydrochloride
1g
C12H19BClNO2
2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline hydrochloride, 2-aminophenylboronic acid, pinacol ester, hcl, 2-tetramethyl-1,3,2-dioxaborolan-2-yl aniline hydrochloride, 2-aminobenzeneboronic acid pinacol ester hci, 2-aminophenyl boronic acid, pinacol ester hydrochloride, 2-aminophenyl boronic acid,pinacol ester hydrochloride, 2-aminobenzeneboronic acid pinacol ester hydrochloride, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline-hydrogen chloride 1/1, benzenamine, 2-4,4,5,5-tetremethyl-1,3,2-dioxaborolan-2-yl-, hcl salt
B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2N.Cl
255.549
255.55
161°C to 163°C
393877-09-3
MFCD02179449
XWILEMYKORLIPW-UHFFFAOYSA-N
2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline;hydrochloride
44118691
96%

Chemical Identifiers

393877-09-3
255.549
XWILEMYKORLIPW-UHFFFAOYSA-N
44118691
B1(OC(C(O1)(C)C)(C)C)C2=CC=CC=C2N.Cl
C12H19BClNO2
MFCD02179449
2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline hydrochloride, 2-aminophenylboronic acid, pinacol ester, hcl, 2-tetramethyl-1,3,2-dioxaborolan-2-yl aniline hydrochloride, 2-aminobenzeneboronic acid pinacol ester hci, 2-aminophenyl boronic acid, pinacol ester hydrochloride, 2-aminophenyl boronic acid,pinacol ester hydrochloride, 2-aminobenzeneboronic acid pinacol ester hydrochloride, 2-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline-hydrogen chloride 1/1, benzenamine, 2-4,4,5,5-tetremethyl-1,3,2-dioxaborolan-2-yl-, hcl salt
2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents