Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Aminobenzanilide, 95%

$39.88 - $143.78

Chemical Identifiers

Chemical Name or Material 2-Aminobenzanilide
CAS 4424-17-3
Molecular Formula C13H12N2O
Formula Weight 212.25
MDL Number MFCD00034792
Beilstein 782786
Synonym 2-Amino-N-phenylbenzamide; Phenylanthranilamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1145106

  SDS
alfa aesar™
L1145106
5g Each for $39.88

AAL1145114

  SDS
alfa aesar™
L1145114
25g Each for $143.78
Description & Specifications

Specifications

Formula Weight 212.25
Chemical Name or Material 2-Aminobenzanilide
Assay 95%
Molecular Formula C13H12N2O
MDL Number MFCD00034792
CAS 4424-17-3
Beilstein 782786
Synonym 2-Amino-N-phenylbenzamide; Phenylanthranilamide
Melting Point 130° to 134°C
Quantity 5g
RTECS Number CV2325000