missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Aminobenzanilide, 95%

Manufacturer:  Alfa Aesar™ L1145114

 View more versions of this product

Catalog No. AAL1145114



Specifications

Specifications

2-Aminobenzanilide
25g
C13H12N2O
782786
FDPVTENMNDHFNK-UHFFFAOYSA-N
2-amino-N-phenylbenzamide
78142
95%
130°C to 134°C
4424-17-3
MFCD00034792
2-aminobenzanilide, benzamide, 2-amino-n-phenyl, anthranilanilide, n-2-aminobenzoyl aniline, n-phenylanthranilamide, benzanilide, 2-amino, n-phenyl-2-aminobenzamide, 2-amino-n-phenyl-benzamide, 2-aminophenyl-n-benzamide, o-aminobenzanilide
C1=CC=C(C=C1)NC(=O)C2=CC=CC=C2N
212.252
212.25

Chemical Identifiers

4424-17-3
212.252
FDPVTENMNDHFNK-UHFFFAOYSA-N
78142
C1=CC=C(C=C1)NC(=O)C2=CC=CC=C2N
C13H12N2O
MFCD00034792
2-aminobenzanilide, benzamide, 2-amino-n-phenyl, anthranilanilide, n-2-aminobenzoyl aniline, n-phenylanthranilamide, benzanilide, 2-amino, n-phenyl-2-aminobenzamide, 2-amino-n-phenyl-benzamide, 2-aminophenyl-n-benzamide, o-aminobenzanilide
2-amino-N-phenylbenzamide
Safety and Handling

Safety and Handling

RTECSNumber : CV2325000

Documents