missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Aminobenzamidoxime, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H5178203

 View more versions of this product

Catalog No. AAH5178203



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Aminobenzamidoxime
71°C to 75°C
MFCD00492723
2-aminobenzamide oxime, 2-aminobenzamidoxime, 2-amino-n'-hydroxybenzimidamide, 2-amino-n-hydroxybenzimidamide, z-2-amino-n'-hydroxybenzimidamide, e-2-amino-n'-hydroxybenzenecarboximidamide
C1=CC=C(C(=C1)C(=NO)N)N
151.169
151.17
16348-49-5
C7H9N3O
1 g
CFZHYRNQLHEHJS-UHFFFAOYSA-N
2-amino-N'-hydroxybenzenecarboximidamide
9628840
97%

Chemical Identifiers

16348-49-5
151.169
CFZHYRNQLHEHJS-UHFFFAOYSA-N
9628840
C1=CC=C(C(=C1)C(=NO)N)N
C7H9N3O
MFCD00492723
2-aminobenzamide oxime, 2-aminobenzamidoxime, 2-amino-n'-hydroxybenzimidamide, 2-amino-n-hydroxybenzimidamide, z-2-amino-n'-hydroxybenzimidamide, e-2-amino-n'-hydroxybenzenecarboximidamide
2-amino-N'-hydroxybenzenecarboximidamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only