missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Aminobenzamide, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1475622

 View more versions of this product

Catalog No. AAA1475622



Description

Description

2-Aminobenzamide is used as an acetyldehyde scavenger in PET bottles. It is also used as an intermediate for ag chemicals, dyes, pharmaceuticals, blowing agents, and fine organic chemicals. In lubricating oils for supersonic jet engines, inhibits copper and magnesium corrosion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Aminobenzamide is used as an acetyldehyde scavenger in PET bottles. It is also used as an intermediate for ag chemicals, dyes, pharmaceuticals, blowing agents, and fine organic chemicals. In lubricating oils for supersonic jet engines, inhibits copper and magnesium corrosion.

Solubility
Soluble in hot water and alcohol. Slightly soluble in ether and benzene. Very soluble in ethyl acetate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Specifications

2-Aminobenzamide
110°C to 114°C
198°C (388°F)
MFCD00007981
508509
Soluble in hot water and alcohol. Slightly soluble in ether and benzene. Very soluble in ethyl acetate.
C1=CC=C(C(=C1)C(=O)N)N
136.154
136.15
88-68-6
300°C
C7H8N2O
100 g
anthranilamide, benzamide, 2-amino, o-aminobenzamide, 2-carbamoylaniline, aminobenzamide, anthranilimidic acid, benzamide, o-amino, anthranilic acid amide, anthranilamide van, o-aminobenzamide van
PXBFMLJZNCDSMP-UHFFFAOYSA-N
2-aminobenzamide
6942
≥98%

Chemical Identifiers

88-68-6
136.154
PXBFMLJZNCDSMP-UHFFFAOYSA-N
6942
C1=CC=C(C(=C1)C(=O)N)N
C7H8N2O
MFCD00007981
anthranilamide, benzamide, 2-amino, o-aminobenzamide, 2-carbamoylaniline, aminobenzamide, anthranilimidic acid, benzamide, o-amino, anthranilic acid amide, anthranilamide van, o-aminobenzamide van
2-aminobenzamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P280-P305+P351+P338-P310

H318

EINECSNumber : 201-851-2

RTECSNumber : CU8993000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only