Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Aminoanthraquinone, tech. 90%

$118.99 - $118.99

Chemical Identifiers

Chemical Name or Material 2-Aminoanthraquinone
CAS 117-79-3
Purity Grade Technical Grade
Molecular Formula C14H9NO2
Formula Weight 223.23
MDL Number MFCD00001233
Beilstein 745903
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0336836

 
alfa aesar™
L0336836
500g Each for $118.99
Description & Specifications

Specifications

Formula Weight 223.23
Chemical Name or Material 2-Aminoanthraquinone
Molecular Formula C14H9NO2
MDL Number MFCD00001233
Health Hazard 2 GHS H Statement
H351
Suspected of causing cancer.
Odor Odorless
TSCA TSCA
Purity Grade Technical Grade
Assay 90%
CAS 117-79-3
Beilstein 745903
Melting Point ≈293°C
Quantity 500g
Flash Point 283°C (541°F)
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P280h
Wear protective gloves/clothing.
RTECS Number CB5120000
Safety and Handling

Warning

GHS P Statement
P280h
Wear protective gloves/clothing.

GHS H Statement
H351
Suspected of causing cancer.