missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Aminoadenosine, 97%

Manufacturer:  Alfa Aesar™ 4474803

 View more versions of this product

Catalog No. AA4474803



Specifications

Specifications

2-Aminoadenosine
C6H5ClFN
benzenamine, 2-chloro-4-fluoro, 2-chloro-4-fluorobenzenamine, 2-chloro-4-fluorophenylamine, 2-chloro-4-fluoro-phenylamine, pubchem2943, 2-chloro-4fluoroaniline, 2-chloro-4-fluroaniline, timtec-bb sbb004110, 4-fluoro-2-chloroaniline, acmc-1cj1h
C1=CC(=C(C=C1F)Cl)N
145.56
282.26
97%
2106-02-7
MFCD00053556
XRAKCYJTJGTSMM-UHFFFAOYSA-N
2-chloro-4-fluoroaniline
75013
Powder
1g

Chemical Identifiers

2106-02-7
145.56
XRAKCYJTJGTSMM-UHFFFAOYSA-N
75013
C1=CC(=C(C=C1F)Cl)N
C6H5ClFN
MFCD00053556
benzenamine, 2-chloro-4-fluoro, 2-chloro-4-fluorobenzenamine, 2-chloro-4-fluorophenylamine, 2-chloro-4-fluoro-phenylamine, pubchem2943, 2-chloro-4fluoroaniline, 2-chloro-4-fluroaniline, timtec-bb sbb004110, 4-fluoro-2-chloroaniline, acmc-1cj1h
2-chloro-4-fluoroaniline
SDS Documents