missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Aminoadenosine, 97%

Manufacturer:  Alfa Aesar™ 4474806

 View more versions of this product

Catalog No. AA4474806



Specifications

Specifications

2-Aminoadenosine
C10H14N6O4
2-aminoadenosine, 2-amine adenosine, adenosine, 2-amino, 2,6-diaminopurine riboside, 2,6-diaminonebularine, 2,6-diaminopurinosine, 2r,3r,4s,5r-2-2,6-diaminopurin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, 2,6-diamino-9-beta-d-ribofuranosyl purine, pubchem14156, ksc202m1p
C1=NC2=C(N1C3C(C(C(O3)CO)O)O)N=C(N=C2N)N
282.26
CHEBI:1014
Powder
5g
2096-10-8
MFCD00053556
ZDTFMPXQUSBYRL-UHFFFAOYNA-N
(2R,3R,4S,5R)-2-(2,6-diaminopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
72200
282.26
97%

Chemical Identifiers

2096-10-8
282.26
ZDTFMPXQUSBYRL-UHFFFAOYNA-N
72200
(2R,3R,4S,5R)-2-(2,6-diaminopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
C10H14N6O4
MFCD00053556
2-aminoadenosine, 2-amine adenosine, adenosine, 2-amino, 2,6-diaminopurine riboside, 2,6-diaminonebularine, 2,6-diaminopurinosine, 2r,3r,4s,5r-2-2,6-diaminopurin-9-yl-5-hydroxymethyl tetrahydrofuran-3,4-diol, 2,6-diamino-9-beta-d-ribofuranosyl purine, pubchem14156, ksc202m1p
CHEBI:1014
C1=NC2=C(N1C3C(C(C(O3)CO)O)O)N=C(N=C2N)N
SDS Documents