missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-p-cresol, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC163210010

 View more versions of this product

Catalog No. AC163210010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Amino-p-cresol
Glass bottle
134.0°C to 139.0°C
97%
96.0
96% min. (GC)
H2NC6H3(CH3)OH
13,601
Solubility in water: slightly soluble
CC1=CC(=C(C=C1)O)N
123.16
123.16
97%
Authentic
Beige to Brown-Gray
1kg
95-84-1
100.0
C7H9NO
MFCD00007699
2-amino-4-cresol
ZMXYNJXDULEQCK-UHFFFAOYSA-N
2-amino-4-methylphenol
7264
Flakes or Crystalline Powder

Chemical Identifiers

95-84-1
123.16
ZMXYNJXDULEQCK-UHFFFAOYSA-N
7264
CC1=CC(=C(C=C1)O)N
C7H9NO
MFCD00007699
2-amino-4-cresol
2-amino-4-methylphenol
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-457-3

RTECSNumber : SJ6078000

TSCA : TSCA

SDS Documents