Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-N-sec-butylbenzamide, 96%

$147.92 - $147.92

Chemical Identifiers

Chemical Name or Material 2-Amino-N-sec-butylbenzamide
CAS 30391-87-8
Molecular Formula C11H16N2O
Formula Weight 192.26
MDL Number MFCD00116303
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5072403

  SDS
alfa aesar™
H5072403
1g Each for $147.92
Description & Specifications

Specifications

Formula Weight 192.26
Chemical Name or Material 2-Amino-N-sec-butylbenzamide
Assay 96%
Quantity 1g
Molecular Formula C11H16N2O
MDL Number MFCD00116303
CAS 30391-87-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.