missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-9-fluorenone, 98%

Manufacturer:  Alfa Aesar™ H5504814

 View more versions of this product

Catalog No. AAH5504814



Description

Description

Applications
2-Amino-9-fluorenone is used as an active pharmaceutical intermediate.

Solubility
Soluble in alcohol and ether. Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

2-Amino-9-fluorenone
25g
C13H9NO
2-amino-9-fluorenone
SJODITPGMMSNRF-UHFFFAOYSA-N
2-aminofluoren-9-one
18357
98%
157°C to 160°C
3096-57-9
MFCD00001157
Soluble in alcohol and ether. Insoluble in water.
C1=CC=C2C(=C1)C3=C(C2=O)C=C(C=C3)N
195.221
195.22

Chemical Identifiers

3096-57-9
195.221
SJODITPGMMSNRF-UHFFFAOYSA-N
18357
C1=CC=C2C(=C1)C3=C(C2=O)C=C(C=C3)N
C13H9NO
MFCD00001157
2-amino-9-fluorenone
2-aminofluoren-9-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 221-445-9

RTECSNumber : LL8980200

TSCA : No

Recommended Storage : Ambient temperatures

Documents