missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-7-bromofluorene, tech. 90%

Manufacturer:  Alfa Aesar™ H55370MD

 View more versions of this product

Catalog No. AAH55370MD



Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Sparingly soluble in water. (0.724 mg/L)

Notes
Incompatible with oxidizing agents. Store at room temperature.
Specifications

Specifications

2-Amino-7-bromofluorene
250mg
C13H10BrN
2-amino-7-bromo-9h-fluorene
RJWYTISHBMNMOZ-UHFFFAOYSA-N
7-bromo-9H-fluoren-2-amine
23126
90%
148°C to 149°C
6638-60-4
MFCD00001127
Sparingly soluble in water. (0.724mg/L)
C1C2=C(C=CC(=C2)N)C3=C1C=C(C=C3)Br
260.134
260.13
Technical

Chemical Identifiers

6638-60-4
260.134
RJWYTISHBMNMOZ-UHFFFAOYSA-N
23126
C1C2=C(C=CC(=C2)N)C3=C1C=C(C=C3)Br
C13H10BrN
MFCD00001127
2-amino-7-bromo-9h-fluorene
7-bromo-9H-fluoren-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

RTECSNumber : LL5150050

TSCA : No

Recommended Storage : Ambient temperatures

Documents