Learn More
2-Amino-7-bromofluorene, tech. 90%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H5537006
Description
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Sparingly soluble in water. (0.724 mg/L)
Notes
Incompatible with oxidizing agents. Store at room temperature.
Specifications
2-Amino-7-bromofluorene | |
148°C to 149°C | |
MFCD00001127 | |
2-amino-7-bromofluorene, 2-bromo-7-aminofluorene, fluoren-2-amine, 7-bromo, 7-bromo-9h-fluoren-2-ylamine, 9h-fluoren-2-amine, 7-bromo, 7-bromofluorene-2-ylamine, acmc-20a5iw, 2-amino-7-bromo-9h-fluorene, 9h-fluoren-2-amine,7-bromo, 7-bromanyl-9h-fluoren-2-amine | |
RJWYTISHBMNMOZ-UHFFFAOYSA-N | |
7-bromo-9H-fluoren-2-amine | |
23126 | |
90% |
6638-60-4 | |
C13H10BrN | |
5 g | |
Sparingly soluble in water. (0.724mg/L) | |
C1C2=C(C=CC(=C2)N)C3=C1C=C(C=C3)Br | |
260.134 | |
260.13 | |
Technical |
Chemical Identifiers
6638-60-4 | |
260.134 | |
RJWYTISHBMNMOZ-UHFFFAOYSA-N | |
23126 | |
C1C2=C(C=CC(=C2)N)C3=C1C=C(C=C3)Br |
C13H10BrN | |
MFCD00001127 | |
2-amino-7-bromofluorene, 2-bromo-7-aminofluorene, fluoren-2-amine, 7-bromo, 7-bromo-9h-fluoren-2-ylamine, 9h-fluoren-2-amine, 7-bromo, 7-bromofluorene-2-ylamine, acmc-20a5iw, 2-amino-7-bromo-9h-fluorene, 9h-fluoren-2-amine,7-bromo, 7-bromanyl-9h-fluoren-2-amine | |
7-bromo-9H-fluoren-2-amine |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
RTECSNumber : LL5150050
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only