missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Amino-7-bromofluorene, tech. 90%

Manufacturer:  Alfa Aesar™ H5537003

 View more versions of this product

Catalog No. AAH5537003



Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Sparingly soluble in water. (0.724 mg/L)

Notes
Incompatible with oxidizing agents. Store at room temperature.
Specifications

Specifications

2-Amino-7-bromofluorene
1g
C13H10BrN
2-amino-7-bromo-9h-fluorene
C1C2=C(C=CC(=C2)N)C3=C1C=C(C=C3)Br
260.134
260.13
Technical
148°C to 149°C
6638-60-4
MFCD00001127
RJWYTISHBMNMOZ-UHFFFAOYSA-N
7-bromo-9H-fluoren-2-amine
23126
90%

Chemical Identifiers

6638-60-4
260.134
RJWYTISHBMNMOZ-UHFFFAOYSA-N
23126
C1C2=C(C=CC(=C2)N)C3=C1C=C(C=C3)Br
C13H10BrN
MFCD00001127
2-amino-7-bromo-9h-fluorene
7-bromo-9H-fluoren-2-amine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : LL5150050

Documents