Please login to your online account to display your discounted pricing

2-Amino-6-purinethiol, 95%, ACROS Organics™


Quantity 250mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC104482500

 View more versions of this product

Catalog No. AC104482500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Color Yellow to green
Physical Form Powder
Formula Weight 167.19
Quantity 250mg
Beilstein 26, IV, 3926
Merck Index 15, 9490
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 2-Amino-6-purinethiol
Molecular Formula C5H5N5S
MDL Number MFCD00127785
Health Hazard 1 Danger
Packaging Glass bottle
Synonym 6-Thioguanine
Infrared Spectrum Authentic
Assay Percent Range 95%
CAS 154-42-7
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Toxic if swallowed.

Specifications

Chemical Name or Material 2-Amino-6-purinethiol
CAS 154-42-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C5H5N5S
Formula Weight 167.19
MDL Number MFCD00127785
Synonym 6-Thioguanine
Accessories

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.

SDS & Safety Documents