Please login to your online account to display your discounted pricing

2-Amino-6-purinethiol, 95%, ACROS Organics™

$168.03 - $168.03

Chemical Identifiers

Chemical Name or Material 2-Amino-6-purinethiol
CAS 154-42-7
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C5H5N5S
Formula Weight 167.19
MDL Number MFCD00127785
Synonym 6-Thioguanine
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC104482500

  SDS
thermo fisher scientific
AC104482500
Glass bottle 250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC104480010

  SDS
thermo fisher scientific
104480010
Glass bottle 1g Each for $168.03

AC104480050

  SDS
thermo fisher scientific
104480050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Color Yellow to green
Physical Form Powder
Formula Weight 167.19
Quantity 250mg
Beilstein 26, IV, 3926
Merck Index 15, 9490
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 2-Amino-6-purinethiol
Assay Percent Range 95%
Molecular Formula C5H5N5S
MDL Number MFCD00127785
CAS 154-42-7
Infrared Spectrum Authentic
Synonym 6-Thioguanine
Health Hazard 1 Danger
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.