missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Amino-6-purinethiol, 95%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 104480050

 View more versions of this product

Catalog No. AC104480050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-Amino-6-purinethiol
Authentic
100.0
Yellow to green
5g
95%
167.19
26, IV, 3926
6-Thioguanine
154-42-7
94.0
Glass bottle
Powder
≥94%
C5H5N5S
MFCD00127785
15, 9490

Chemical Identifiers

154-42-7
100.0
C5H5N5S
MFCD00127785
2-Amino-6-purinethiol
94.0
95%
167.19
6-Thioguanine
≥94%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Toxic if swallowed.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger